Skip to Content
Merck
All Photos(1)

Documents

PHL84053

Artepillin C

phyproof® Reference Substance

Synonym(s):

(2E)-3-[4-Hydroxy-3,5-bis(3-methyl-2-buten-1-yl)phenyl]-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

InChI

1S/C19H24O3/c1-13(2)5-8-16-11-15(7-10-18(20)21)12-17(19(16)22)9-6-14(3)4/h5-7,10-12,22H,8-9H2,1-4H3,(H,20,21)/b10-7+

InChI key

KABCFARPAMSXCC-JXMROGBWSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • Antineoplastic activity in colorectal cancer models: Research highlighted the antineoplastic properties of Artepillin C and Brazilian brown propolis in colorectal areas of Wistar rats, suggesting its potential application in cancer therapy (Pelegrini et al., 2024).
  • pH-dependent cytotoxicity against tumor cells: A study examined the pH-dependent cytotoxic effects of Artepillin C on tumor cells, providing insights into its therapeutic potential for cancer treatment (Pazin et al., 2023).
  • Antitumor potential against breast cancer: Artepillin C demonstrated significant antitumor potential against breast cancer cell lines, indicating its relevance in developing new anticancer therapies (de Freitas Meirelles et al., 2024).
  • Potential anti-obesogenic properties: Artepillin C is reviewed for its potential as an anti-obesogenic agent, exploring its use in obesity prevention and management strategies (Tek et al., 2024).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service