Skip to Content
Merck
All Photos(1)

Documents

C8221

Sigma-Aldrich

Caffeic acid phenethyl ester

≥97% (HPLC), powder

Synonym(s):

CAPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16O4
CAS Number:
Molecular Weight:
284.31
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥97% (HPLC)

form

powder

storage condition

protect from light

color

off-white

solubility

ethyl acetate: 50 mg/mL
DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

Oc1ccc(\C=C\C(=O)OCCc2ccccc2)cc1O

InChI

1S/C17H16O4/c18-15-8-6-14(12-16(15)19)7-9-17(20)21-11-10-13-4-2-1-3-5-13/h1-9,12,18-19H,10-11H2/b9-7+

InChI key

SWUARLUWKZWEBQ-VQHVLOKHSA-N

Gene Information

human ... NFKB2(4791)

Application

Caffeic acid phenethyl ester (CAPE) has been used to determine the effect of CAPE in hyperthyroidism, as a component of culture media to measure the cytotoxicity of breast cancer cell line MDA-MB-231, to study its effects on amiodarone-induced pulmonary fibrosis in rats.

Biochem/physiol Actions

CAPE exhibits several properties such as anticarcinogenic, immunomodulatory activities, antioxidant, antiviral, anti-inflammatory, hypoglycaemic effect, cardioprotective and inhibits gastric secretion. It also has an inhibitory effect on fibroblasts growth factor type 2 (FGF-2), a tumor growth factor in breast cancer.
Caffeic acid phenethyl ester is a specific inhibitor of the nuclear transcription factor NF-κB.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z S Zheng et al.
Oncology research, 7(9), 445-452 (1995-01-01)
Caffeic acid phenethyl ester (CAPE) was evaluated for its potential in regulating keratinocyte proliferation. CAPE inhibited the proliferation of SV40 transformed keratinocytes (Z114) in a concentration- and time-dependent manner. Inhibition by CAPE was seen with 0.5 to 5.0 micrograms/ml at
M R Fesen et al.
Biochemical pharmacology, 48(3), 595-608 (1994-08-03)
The inhibition of HIV-1 integrase by flavones and related compounds was investigated biochemically and by means of structure-activity relationships. Purified enzyme and synthetic oligonucleotides were used to assay for three reactions catalysed by integrase: (1) processing of 3' termini by
D Grunberger et al.
Experientia, 44(3), 230-232 (1988-03-15)
The honeybee hive product, propolis, is a folk medicine employed for treating various ailments. Many important pharmaceutical properties have been ascribed to propolis, including anti-inflammatory, antiviral, immunostimulatory and carcinostatic activities. Propolis extracts have provided an active component identified as caffeic
G F Sud'ina et al.
FEBS letters, 329(1-2), 21-24 (1993-08-23)
Caffeic acid phenethyl ester, an active component of propolis extract, inhibits 5-lipoxygenase in the micromolar concentration range. The inhibition is of an uncompetitive type, i.e. the inhibitor binds to the enzyme-substrate complex but not to the free enzyme. Caffeic acid
Z Z Su et al.
Anticancer research, 15(5B), 1841-1848 (1995-09-01)
The active component of the folk medicine propolis, caffeic acid phenethyl ester (CAPE), displays selective toxicity toward cloned rat embryo fibroblast (CREF) cells transformed by a spectrum of diverse acting oncogenes. Identification of the mode of action of CAPE should

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service