Skip to Content
Merck
All Photos(1)

Key Documents

CRM48620

Supelco

Toluene solution

certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL

Synonym(s):

Toluene solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tin-Tin Win-Shwe et al.
Toxicology letters, 198(2), 93-99 (2010-07-06)
Chemical susceptibility is triggered by a large range of chemicals present both indoors and outdoors including pesticides, cleaning products, perfumes, scented products and cigarette smoke. Health risk after chemical exposure depends on age, sex, genetic factors, socioeconomic status, nutritional status
Andreas Seidler et al.
Occupational and environmental medicine, 71(4), 259-265 (2014-02-04)
To evaluate the association between dinitrotoluene (DNT) exposure and renal cancer in a case-cohort study. This case-cohort study was conducted among men born between 1920 and 1974 (n=16 441) who were gainfully employed between 1953 and 1990 in one of
Luis Cuauhtémoc Haro-García et al.
Revista medica del Instituto Mexicano del Seguro Social, 46(6), 643-650 (2009-03-07)
Despite, the idea promoted to study occupational exposure to benzene and its mixture with toluene and xylene (BTX) because it appears to determine its toxicity and probably the production of additive effects, it persists interest to recognizing its hematological and
John H Hannigan et al.
Systems biology in reproductive medicine, 56(2), 184-200 (2010-04-10)
Toluene is an organic solvent that is widely used by industry and is ubiquitous in our environment. As a result, exposure to solvents like toluene in work-related settings (i.e., relatively constant, low-level exposures) or through inhalant abuse (i.e., relatively intermittent
[Advance on biological monitoring of occupational exposure to toluene].
Hong-xia Chen et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 28(8), 613-615 (2010-12-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service