CRM44895
ASTM® D6584 Triolein solution
certified reference material, 5000 μg/mL in pyridine, ampule of 1 × 2 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
Agency
ASTM® D6584
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
ampule of 1 × 2 mL
concentration
5000 μg/mL in pyridine
application(s)
petroleum
format
single component solution
storage temp.
2-8°C
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
62.6 °F - closed cup
Flash Point(C)
17.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service