Skip to Content
Merck
All Photos(1)

Key Documents

CRM01373

Supelco

Tebufenozide

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O2
CAS Number:
Molecular Weight:
352.47
Beilstein:
7822297
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

CCc1ccc(cc1)C(=O)NN(C(=O)c2cc(C)cc(C)c2)C(C)(C)C

InChI

1S/C22H28N2O2/c1-7-17-8-10-18(11-9-17)20(25)23-24(22(4,5)6)21(26)19-13-15(2)12-16(3)14-19/h8-14H,7H2,1-6H3,(H,23,25)

InChI key

QYPNKSZPJQQLRK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Tebufenozide is a non-steroidal ecdysone agonist herbicide that belongs to the class of diacylhydrazines. It interacts with the steroidal receptors in lepidopterans larvae and produces lethal molting.
Tebufenozide was allowed for use in the European Union as a plant protection agent as per the Commission Implementing Directive 2011/60/EU of 23rd May 201, repealing Directive 91/414/EEC. A maximum residue limit (MRL) of 2 mg/kg and 0.05 mg/kg is set for the presence of tebufenozide in fresh/frozen fruits and tree nuts respectively according to the Reg. (EU) 2021/1807.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
Tebufenozide certified reference material (CRM) may also find the following uses:

  • Analysis of trifloxystrobin, tebufenozide, and halofenozide by Micellar electrokinetic capillary chromatography (MEKC) in tomato, celery, and apple juice samples
  • Measurement of the residue levels of tebufenozide in cabbage and soil by reversed-phase high-performance liquid chromatography (HPLC)
  • Determination of four diacylhydrazine insecticide residues in apple, grape, cucumber, tomato, and spinach samples by QuEChERS method using ultra-performance liquid chromatography coupled with tandem mass spectrometry
  • Development and validation of a multi-residual analytical method, based on SPE followed by LC-MS for the simultaneous estimation of 14 pesticides in surface and groundwaters
  • Multi-residue analysis of 200 pesticides and pesticide metabolites in honeybees by gas and liquid chromatography coupled with tandem mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Liu et al.
Ecotoxicology and environmental safety, 134P1, 204-212 (2016-09-17)
The novel nonsteroidal ecdysone agonist tebufenozide is capable of controlling a wide range of lepidopteran pests. However its intensive use in vegetables has raised concerns towards the safety of food and environment. Here, we developed an easy and reliable method
You Zhou et al.
Ecotoxicology and environmental safety, 189, 110020-110020 (2019-12-07)
Azadirachtin, as the most promising and effective botanical insecticide, exhibits significant growth inhibition activity against agricultural and forestry pests. However, its biochemical effects at the metabolic level compared with those of other insect growth regulators have not been studied. Therefore
Muhammad Fiaz et al.
Chemosphere, 212, 337-345 (2018-08-27)
The velvetbean caterpillar, Anticarsia gemmatalis Hübner (Lepidoptera: Noctuidae), is an important soybean pest in the Americas. Tebufenozide, a novel nonsteroidal ecdysone agonist is used to control this pest. Bioassays were conducted to assess tebufenozide toxicity and their ultrastructural effects on
Determination of tebufenozide and methoxyfenozide residues in vegetables using liquid chromatography-electrospray tandem mass spectrometry
Lin D, et al.
Chinese Journal of Analytical Chemistry, 87-90 null
Xingang Liu et al.
Analytical and bioanalytical chemistry, 401(3), 1051-1058 (2011-06-11)
The new analytical method using Quick, Easy, Cheap, Effective, Rugged, and Safe (QuEChERS) procedure for simultaneous determination of diacylhydrazine insecticide residues in fruits and vegetables was developed using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS). The four insecticides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service