Skip to Content
Merck
All Photos(1)

Key Documents

BP1101

2-[4-[(7-Chloro-4-quinolinyl)amino]pentyl] amino ethanol

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Hydroxychloroquine sulfate, 7-Chloro-4-[4-(N-ethyl-N-b-hydroxyethylamino)-1-methylbutylamino]quinoline sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26ClN3O · H2SO4
CAS Number:
Molecular Weight:
433.95
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

Agency

BP

API family

hydroxychloroquine

form

solid

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 25 mg

manufacturer/tradename

BP

storage condition

OK to freeze

color

off-white

mp

238-241 °C

solubility

water: soluble

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CCN(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12

InChI

1S/C18H26ClN3O.H2O4S/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;1-5(2,3)4/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21);(H2,1,2,3,4)

InChI key

JCBIVZZPXRZKTI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2-[4-[(7-Chloro-4-quinolinyl)amino]pentyl] amino ethanol (Hydroxychloroquine Sulfate) is a salt of hydroxychloroquine (HCQ, Plaquenil), a 4-aminoquinoline based antiviral drug.[1][2][3][4][4][5]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

2-[4-[(7-Chloro-4-quinolinyl)amino]pentyl] amino ethanol BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Biochem/physiol Actions

Hydroxychloroquine sulfate is an anti-inflammatory (via TNF), disease-modifying antirheumatic drug (DMARD), and a heme polymerase inhibitor.
Hydroxychloroquine sulfate is an anti-inflammatory (via TNF), disease-modifying antirheumatic drug (DMARD), and a heme polymerase inhibitor. It is used in treating malaria.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seminars in Arthritis and Rheumatism, 43(2), 264-272 (2013)
Hydroxychloroquine Sulfate
British Pharmacopeia, 2021
British Pharmacopoeia, 44(12), 1000-1004 (2021)
A rapid and reliable method for the quantitation of hydroxychloroquine in serum using turbulent flow liquid chromatography-tandem mass spectrometry
Fuzery AK, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 421 (2013)
Development of a novel ion-pairing HPLC-FL method for the separation and quantification of hydroxychloroquine and its metabolites in whole blood
Charlier B,et al.
Biomedical Chromatography, 32(8), e4258-e4258 (2018)
Pharmacokinetics and bioequivalence study of hydroxychloroquine sulfate tablets in Chinese healthy volunteers by LC--MS/MS
Fan HW, et al.
Rheumatology and therapy, 2(2) (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service