Key Documents
57028
Supelco
Visiprep Manifold Flow Control Valve
for use with DL (Disposable Liner) 12- and 24-port models, pk of 2
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
product line
Visiprep
packaging
pk of 2
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with DL (Disposable Liner) 12- and 24-port models
General description
Manifold flow control valve are disposable liner flow control valves that are used as an accessory for 12 port or 24 port Visiprep manifolds.
Application
Solid-phase extraction (SPE) experiments were performed using a Visiprep DL disposable-liner solid-phase extraction vacuum manifold with individual flow control valves from Supelco (Bellefonte, PA, USA) which enabled twelve SPE tubes to be dried at a time.[1]
Visiprep manifold flow control valve may also be used for solid phase extraction experiments(SPE) in the determination of biogenic amines in wine samples using high-performance liquid chromatography coupled with fluorescence detector (HPLC).[1]
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Determination of biogenic amines in wines by high-performance liquid chromatography with on-column fluorescence derivatization.
Busto O, et al.
Journal of Chromatography A, 757(1-2), 311-318 (1997)