Skip to Content
Merck
All Photos(1)

Key Documents

52665-U

Supelco

Discovery® DSC-SAX SPE Tube

bed wt. 500 mg, volume 6 mL, pk of 30

Synonym(s):

SAX SPE cartridge tube, Strong Anion Exchange

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

polypropylene tube

product line

Discovery®

composition

bed wt., 500 mg

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

surface area

480 m2/g

volume

6 mL

matrix

Silica gel base material (irregularly shaped, acid washed)

matrix active group

ammonium ion, quaternary bonding, polymerically bonded, chloride counter-ion

particle size

50 μm

pore size

0.9 mL/g pore volume
70 Å pore diameter

capacity

0.14 meq/g total capacity

application(s)

food and beverages

separation technique

ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • A polymerically bonded quarternary amine that remains positively charged at all pH levels
  • Counter ion is Cl-
  • Ion exchange capacity is ∼ 0.14 meq/g
  • Commonly used when extracting weaker anions (e.g., carboxylic acids) that may not bind strongly enough to weaker anion-exchangers
  • Selectivity can be modified by changing the counter ion with the appropriate buffer during conditioning

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>464.0 °F

Flash Point(C)

> 240 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service