Skip to Content
Merck
All Photos(1)

Key Documents

52577-U

Supelco

Supelclean SAX/PSA SPE Tube

bed B 500 mg (Supelclean PSA SPE), bed A 500 mg (Supelclean LC-SAX SPE), volume 6 mL, pk of 30

Synonym(s):

Mix-Mode Amine SPE cartridge tube, 6 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

PE frit
polypropylene hardware

product line

Supelclean

composition

bed A, 500 mg (Supelclean LC-SAX SPE)
bed B, 500 mg (Supelclean PSA SPE)

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

matrix active group

PSA phase
SAX phase

application(s)

food and beverages

separation technique

ion exchange
normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Normal-phase and anion-exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • Dual layer SPE tube that contains both Supelclean SAX (upper layer) and PSA (lower layer) SPE sorbents (separated by PE frit)
  • Supelclean SAX is a quarternary amine, Cl- counter-ion.
  • Supelclean PSA an ethylenediamine-N-propyl phas that contains both primary and secondary amines.
  • Ideal for removing matrix components (fatty acids, organic acids, polar pigments, and some sugars) when conducting multi-residue pesticide analysis in foods
  • In compliance with the Luke II method which uses SPE to remove matrix interference and enhancement of pesticides from food for GC-ITMS analysis

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system, respiratory tract irritation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service