Skip to Content
Merck
All Photos(1)

Key Documents

509612

Syringes Fitted with Luer Lock Valve (509353)

volume 5 mL, 5MDR-VLLMA-GT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

glass syringe

needle size

23 ga

feature

Pre-fitted with Luer Lock Valve

packaging

pkg of 1 ea

manufacturer/tradename

SGE 008770

volume

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Syringes Fitted with Luer Lock Valve (509353) are:
  • Ideal for corrosive, radioactive, or sterile materials.
  • Removable handle minimizes heat transfer from hand to sample, making the syringe easy to hold.
  • Accurate and reproducible to +/− 1%.
  • Field repairable.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service