26271
SGE Needles for luer lock syringes
needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm, point style, 5
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
material
cone tip, side port stainless steel needle (point style 5)
needle size
23 ga
description
NLL-5/23H
feature
point style 5
packaging
pkg of 2 ea
manufacturer/tradename
SGE 39803
needle L × O.D.
50 mm × 0.63 mm
fitting
luer lock
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54 -54 (2009)
Kevin Robards, P. E. Jackson, Paul A. Haddad
Principles and Practice of Modern Chromatographic Methods, 83-83 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service