Skip to Content
Merck
All Photos(1)

Documents

176605

Sigma-Aldrich

Chlorobenzene-d5

99 atom % D

Synonym(s):

Pentadeuterochlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D5Cl
CAS Number:
Molecular Weight:
117.59
Beilstein:
1866335
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.11

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

GC/MS: suitable
NMR: suitable

refractive index

n20/D 1.522 (lit.)

bp

130-130.5 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(Cl)c([2H])c1[2H]

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

MVPPADPHJFYWMZ-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorobenzene-d5 is a deuterated derivative of chlorobenzene having an isotopic purity of 99atom%D. 1HNMR (Proton Nuclear Magnetic Resonance) and 2HNMR (Deuterium NMR) spectra of chlorobenzene-d5 were studied in liquid crystal solvents to evaluate the quadrupolar coupling constants (DQCC) and the asymmetry parameters (η) for the deuterons. Values reported were as: DQCCortho = 180(2)kHz, ηortho = 0.06(1), DQCCmeta = 174(2)kHz, ηmeta = 0.09(3), DQCCpara= 182(4)kHz, ηPara = 0.06(4)].
Chlorobenzene-d5 may be used as an internal standard for the high molecular weight and low volatility compounds in thermal desorption/gas chromatography/mass spectrometry (TD/GC/MS) analysis.

Application

May be used as an analytical standard

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The vibrational spectra of penta-deutero-chlorobenzene.
Nanney TR, et al.
Spectrochimica Acta , 21(9), 1495-1504 (1965)
Deuterium quadrupole coupling constants in chlorobenzene-4-d1 and 1, 4-diethylnylbenzene-d1, d' 1 in the liquid state.
Assink RA and Jonas J.
Journal of Magnetic Resonance, 4(3), 347-351 (1971)
A study of volatile organic compounds evolved from the decaying human body.
Statheropoulos M, et al.
Forensic Science International, 153(2), 147-155 (2005)
Deuterium Quadrupolar Parameters from 1H and 2H NMR Spectra for Pyridine-d5, Benzonitrile-d5 and Chlorobenzene-d5 Using Liquid Crystal Solvents.
Ambrosetti R, et al.
Zeitschrift fur Naturforschung, A, 41(1-2), 431-435 (1986)
A study of volatile organic compounds evolved in urban waste disposal bins."
Statheropoulos M, et al.
Atmospheric Environment, 39(26), 4639-4645 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service