Skip to Content
Merck
All Photos(1)

Key Documents

P4100000

Pyridoxine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

pyridoxine

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

214-215 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Pyridoxine hydrochloride is a HCl salt of pyridoxine or vitamin B6. It is a water-soluble vitamin that plays an important role in the treatment of roughness, sunburn, acne, and itch of the skin. It can also be employed in the potential management of sideroblastic anemias, depression and other disorders.

Application

Pyridoxine hydrochloride may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and spectrophotometric techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyridoxine hydrochloride
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 12(9), 3451-3452 (2017)
Electrochemical behaviour of pyridoxine hydrochloride (vitamin B6) at carbon paste electrode modified with crown ethers
Desai PB, et al.
Journal of Solid State Electrochemistry, 12(9), 1067-1075 (2008)
Spectral diagnostics of the interaction between pyridoxine hydrochloride and bovine serum albumin in vitro
Zhang H, et al.
Molecular Biology Reports, 35(4), 699-705 (2008)
Spectrophotometric methods for the simultaneous analysis of meclezine hydrochloride and pyridoxine hydrochloride in bulk drug and pharmaceutical formulations
Arayne MS, et al.
Pakistan Journal of Pharmaceutical Sciences, 20(2), 149-156 (2007)
Simultaneous spectrophotometric analysis of a ternary mixture of pharmaceuticals?assay for meclozine hydrochloride, pyridoxine hydrochloride and caffeine
Sharma SC, et al.
Journal of Pharmaceutical and Biomedical Analysis, 7(3), 321-327 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service