Skip to Content
Merck
All Photos(1)

Key Documents

47845

Supelco

Sorbic acid

analytical standard

Synonym(s):

α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E200

grade

analytical standard

vapor pressure

0.01 mmHg ( 20 °C)

Assay

99.7% (HPLC)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

132-135 °C (lit.)

density

1.2 g/cm3 at 20 °C

antibiotic activity spectrum

fungi

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sorbic acid may be used as a reference standard in the detection and analysis of sorbic acid in alcoholic beverages and different food products, using high performance liquid chromatography (HPLC).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of benzoic and sorbic acids in Brazilian food
Tfouni.V.A.S and Toledo.F.C.M
Food Control, 13(2), 117-123 (2002)
Analysis of benzoic acid and sorbic acid in Thai rice wines and distillates by solid-phase sorbent extraction and high-performance liquid chromatography
Techakriengkrai E and Surakarnkul R
J. Food Compos. Anal., 20, 220-225 (2007)
G D Saraiva et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 137, 1409-1416 (2014-10-14)
This work reports a temperature-dependent vibrational spectroscopic study of the sorbic acid (C6H8O2), as well as the mode assignment at ambient conditions, based on the density functional theory. Temperature-dependent vibrational properties have been performed in polycrystalline sorbic acid through both
A Vermeulen et al.
Food microbiology, 32(2), 389-396 (2012-09-19)
The microbial stability of intermediate moisture foods (IMF) is linked with the possible growth of osmophilic yeast and xerophilic moulds. As most of these products have a long shelf life the assessment of the microbial stability is often an important
Phercyles Veiga-Santos et al.
Acta tropica, 125(3), 349-356 (2012-12-12)
The currently available treatments for Chagas disease show limited therapeutic potential and are associated with serious side effects. Our group has been attempting to find alternative drugs isolated from natural products as a potential source of pharmacological agents against Trypanosoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service