Skip to Content
Merck
All Photos(1)

Documents

47763

Supelco

Cholecalciferol (D3)

analytical standard

Synonym(s):

Cholecalciferol, (+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Beilstein:
2339331
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

form

solid

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

83-86 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1

InChI key

QYSXJUFSXHHAJI-YRZJJWOYSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

Cholecalciferol (D3) is a fat soluble vitamin, and is one of the forms of vitamin D, synthesized in human skin on exposure to ultraviolet (UV) radiation from sunlight, or it can also be obtained from food. It is found to be essential in humans for maintaining normal calcium metabolism.

Application

Cholecalciferol (D3) may be used as an analytical reference for the quantification of the analyte in different food matrices using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Vitamin D acts through a receptor that is a member of the ligand-dependent transcription factor superfamily. Modulates the proliferation and differentiation of both normal and cancer cells. Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells. Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of vitamin D3 in milk-based infant formulas by liquid chromatography-tandem mass spectrometry
Kwak M-B, et al.
Food Chemistry, 165(2), 569-574 (2014)
Simultaneous determination of cholecalciferol (vitamin D3) and ergocalciferol (vitamin D2) in foods by selected reaction monitoring
Dimartino G
Journal of AOAC (Association of Official Analytical Chemists) International, 92(2), 511-517 (2009)
Sara S Oberhelman et al.
Mayo Clinic proceedings, 88(12), 1378-1387 (2013-12-03)
To determine whether a single monthly supplement is as effective as a daily maternal supplement in increasing breast milk vitamin D to achieve vitamin D sufficiency in their infants. Forty mothers with exclusively breast-fed infants were randomized to receive oral
Bich Tran et al.
The American journal of clinical nutrition, 99(1), 156-161 (2013-10-11)
Observational data suggested that supplementation with vitamin D could reduce risk of infection, but trial data are inconsistent. We aimed to examine the effect of oral vitamin D supplementation on antibiotic use. We conducted a post hoc analysis of data
Hongliu Ding et al.
Neurology, 81(17), 1531-1537 (2013-09-27)
To conclusively test for a specific association between the biological marker 25-hydroxy-vitamin D3, a transcriptionally active hormone produced in human skin and liver, and the prevalence and severity of Parkinson disease (PD). We used liquid chromatography/tandem mass spectrometry to establish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service