Skip to Content
Merck
All Photos(1)

Documents

C8267

Sigma-Aldrich

Corn oil

delivery vehicle for fat-soluble compounds

Synonym(s):

Maize oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

maize

form

liquid

color

very faintly yellow to yellow

density

0.9 g/mL at 25 °C (lit.)

application(s)

life science and biopharma
sample preparation

functional group

ester

lipid type

oils

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Corn oil, a highly unsaturated vegetable oil extracted from the germ of corn, is a versatile chemical compound extensively used in biochemical research. It can be used as a carrier/delivery vehicle for fat-soluble compounds, including toxins, and as a glidant in excipient research. Corn oil contains high levels of polyunsaturated fats, including omega-6 and omega-3 fatty acids in a ratio of 2:1. Additionally, when combined with surfactants and gel-forming polymers, it is used in formulation research for developing veterinary vaccines.

Application

Corn oil has been used in research applications as delivery vehicle for fat-soluble compounds including toxins. It is also used in the delivery of tamoxifen (TAM) in mice, curcumin in rats and [3H]retinol in mice.
corn oil has been used in research applications as delivery vehicle for fat-soluble compounds including toxins.

Features and Benefits

  • Can be used in Metabolomics, Biochemical and Formulation research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

489.2 °F - open cup

Flash Point(C)

254 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of Repeated Intraperitoneal Injection of Pharmaceutical-grade and Nonpharmaceutical-grade Corn Oil in Female C57BL/6J Mice
Hubbard JS, et al.
Journal of the American Association For Laboratory Animal Science, 56(6), 779-785 (2017)
Shery Park et al.
Stem cells and development, 25(23), 1801-1807 (2016-10-21)
Repair of calvarial bony defects remains challenging for craniofacial surgeons. Injury experiments on animal calvarial bones are widely used to study healing mechanisms and test tissue engineering approaches. Previously, we identified Gli1+ cells within the calvarial sutures as stem cells
Inhibition of hepatic sulfatase-2 In Vivo: A novel strategy to correct diabetic dyslipidemia
Hassing HC, et al.
Hepatology, 55(6), 1746-1753 (2012)
Content of fatty acids in corn (Zea mays L.) oil from Ecuador
Carrillo W, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 10, 150-153 (2017)
Protective effects of curcumin supplementation on intestinal ischemia reperfusion injury
Okudan N, et al.
Phytomedicine, 20(10), 844-848 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service