Skip to Content
Merck
All Photos(1)

Key Documents

46950-U

Supelco

cis-9,cis-12-Octadecadienoic acid methyl ester

certified reference material, 10 mg/mL in heptane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

single component solution

functional group

ester

storage temp.

-10 to -25°C

SMILES string

O(C)C(=O)CCCCCCC\C=C/C\C=C/CCCCC

InChI

1S/C19H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h7-8,10-11H,3-6,9,12-18H2,1-2H3/b8-7-,11-10-

InChI key

WTTJVINHCBCLGX-NQLNTKRDSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumie Saito et al.
Bioscience, biotechnology, and biochemistry, 74(1), 168-174 (2010-01-09)
gamma-Tocopherol was reacted with (E)-4-oxo-2-nonenal (ONE) at 37 degrees C in an acidic acetonitrile solution. The reaction products were isolated by reversed-phase high-performance liquid chromatography and their structures were characterized to be 5-substituted gamma-tocopherols: 5-(1-(furan-2-yl)pentyl)-gamma-tocopherol (1), 3-(1-butyl-4,5,7-trimethyl-7-(4,8,12-trimethyltridecyl)-8,9-dihydro-7H-furo[3,2-f]chromen-2yl)propanal (2), and 1-(1-butyl-4,5,7-trimethyl-7-(4,8,12-trimethyltridecyl)-8,9-dihydro-7H-furo[3,2-f]chromen-2-yl)-4-(furan-2-yl)octan-3-one
Yasutaka Shimotori et al.
Carbohydrate research, 359, 11-17 (2012-08-29)
Various glycosyl ferulates were efficiently synthesized from 2,4,6-tetra-O-acetyl-α-D-glucopyranosyl bromide (TAGB) with amine by amine-promoted glycosylation without using heavy metal. The resulted acetylated glycosyl ferulates with acetoxyl groups at C-2, C-3 and C-4 were regioselectively deacetylated at C-4 and C-6 positions
Vitaly Roginsky
Chemistry and physics of lipids, 163(2), 127-130 (2009-11-03)
The kinetics of the chain free-radical oxidation of cardiolipin (CL), a unique phospholipid containing four linoleate moieties, have been studied for the first time. The technique based on monitoring oxygen consumption by using a Clark electrode was applied to determine
Cheng-Wu Fang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 33(4), 557-559 (2010-09-18)
To study the chemical constituents of Rhizoma of Coniogramme japonica. The Rhizome of C. japonica were extracted with ethanol and the chemical constituents were separated by chromatography technique, and their structures were determined by spectral analysis. Six compounds were isolated
Akio Kadowaki et al.
Chemistry and physics of lipids, 165(2), 178-185 (2012-01-03)
The antioxidative action of fullerenes has received much attention, but their reaction mechanism toward lipid-derived peroxyl radicals has not been well elucidated. In this study, the reaction products of [60]fullerene (C(60)) during the autoxidation of methyl linoleate (MeL) were isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service