Skip to Content
Merck
All Photos(1)

Key Documents

46935-U

Supelco

Isobutyric acid

analytical standard

Synonym(s):

2-Methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2H
CAS Number:
Molecular Weight:
88.11
Beilstein:
635770
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.04 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

824 °F

expl. lim.

10 %

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.393 (lit.)

bp

153-154 °C (lit.)

mp

−47 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CC(C)C(O)=O

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)

InChI key

KQNPFQTWMSNSAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Vyalikh et al.
Physical chemistry chemical physics : PCCP, 9(18), 2249-2257 (2007-05-10)
Solid state deuterium NMR has been used to study the molecular motion of d(6)-isobutyric acid (d(6)-iBA) in the pure (unconfined) state and confined in the cylindrical pores of two periodic mesoporous silica materials (MCM-41, pore size 3.3 nm and SBA-15
E M Faed et al.
Clinical and experimental pharmacology & physiology, 5(2), 195-198 (1978-03-01)
1. Two main conjugates of CPIB (2-[chlorophenoxy]-2-methylpropionic acid) are present in the urine of subjects taking clofibrate. The metabolites can be separated by thin-layer chromatography (TLC). 2. Both conjugates are hydrolysed by dilute alkali, but only one is hydrolysed by
A synthetic metabolic pathway for production of the platform chemical isobutyric acid.
Kechun Zhang et al.
ChemSusChem, 4(8), 1068-1070 (2011-07-12)
Julia Biermann et al.
Molecular vision, 17, 395-403 (2011-02-12)
Histone deacetylase inhibitors (HDACi) have neuroprotective effects under various neurodegenerative conditions, e.g., after optic nerve crush (ONC). HDACi-mediated protection of central neurons by increased histone acetylation has not previously been demonstrated in rat retinal ganglion cells (RGCs), although epigenetic changes
Nicoletta Beffagna et al.
Journal of experimental botany, 58(15-16), 4183-4194 (2007-11-28)
In Arabidopsis thaliana cells, fusicoccin (FC) treatment induced an early and marked increase in the extracellular H(2)O(2) level. It also increased the huge hypo-osmotic stress-induced oxidative wave and, in addition, prevented the H(2)O(2) peak drop. These effects were apparently not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service