Skip to Content
Merck
All Photos(2)

Key Documents

43540

Supelco

Diisobutyl phthalate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

DIBP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2CH2CH(CH3)2]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
2054802
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.49 (lit.)

bp

327 °C (lit.)

density

1.039 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)COC(=O)c1ccccc1C(=O)OCC(C)C

InChI

1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3

InChI key

MGWAVDBGNNKXQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Borch et al.
Toxicology letters, 163(3), 183-190 (2006-02-07)
Phthalates are widely used as plasticizers in various consumer products and building materials. Some of the phthalates are known to interfere with male reproductive development in rats, and di-n-butyl phthalate (DBP), diethylhexyl phthalate (DEHP) and butyl benzyl phthalate (BBP) were
Feng Zeng et al.
Journal of hazardous materials, 164(2-3), 1171-1178 (2008-10-31)
Surface soils from 37 sampling sites including roadsides (RS), parks (PA) and residential areas (RE) of the Subtropical City, Guangzhou, were collected in December of 2005 and analyzed for 16 phthalate esters (PAEs). PAEs were detected in all surface soils
Anne-Marie Saillenfait et al.
Reproductive toxicology (Elmsford, N.Y.), 26(2), 107-115 (2008-08-19)
Diisobutyl phthalate (DIBP) is the branched isomer of DBP; DBP side chains have a four-carbon backbone (C4), whereas DIBP has its four-carbon alkyl side chains rearranged into a three-carbon backbone (C3) with a methyl branch. Di-n-butyl phthalate (DBP), and several
Feng Zeng et al.
Environmental pollution (Barking, Essex : 1987), 156(2), 425-434 (2008-03-18)
This study reports the first data on the concentrations and distribution of phthalate esters (PAEs) in the agricultural soils from the peri-urban areas of Guangzhou city. Sigma(16)PAEs concentrations ranged from 0.195 to 33.6 microg g(-1)-dry weight (dw). Elevated levels of
Ajin V Cheruvathur et al.
Langmuir : the ACS journal of surfaces and colloids, 28(5), 2643-2651 (2012-01-06)
To study the surface structure of MgCl(2) support and its interaction with other active components in Ziegler-Natta catalyst, such as electron donors, we prepared a thin film analogue for Ziegler-Natta ethylene polymerization catalyst support by spin-coating a solution of MgCl(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service