Skip to Content
Merck
All Photos(1)

Key Documents

40027

Supelco

1,1-Dichloroethene solution

certified reference material, 1000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2
CAS Number:
Molecular Weight:
96.94
EC Number:
MDL number:
UNSPSC Code:
41116105
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

ClC(=C)Cl

InChI

1S/C2H2Cl2/c1-2(3)4/h1H2

InChI key

LGXVIGDEPROXKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

1,1-Dichloroethene is a highly reactive and flammable liquid[1], which can play the role of an intermediate in the production of copolymers[2] with vinyl chloride or acrylonitrile.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Larson.AR
Biohazards of Drinking Water Treatment (1988)
Renal tumorigenicity of 1, 1-dichloroethene in mice: the role of male-specific expression of cytochrome P450 2E1 in the renal bioactivation of 1, 1-dichloroethene
Toxicology and Applied Pharmacology, 130(1), 48-56 (1995)
Masarin Ban et al.
Toxicology, 226(2-3), 161-171 (2006-08-02)
Occupational allergy and asthma is a challenging issue in the developing countries. Chemicals inhaled in the workplaces may act not only as allergens but also as immune response modifiers, contributing to asthma exacerbation. In this study, we tested the adjuvant
Daniel B Cope et al.
Environmental science & technology, 43(1), 169-175 (2009-02-13)
A feasibility study was conducted using slags from six grey-iron foundries to evaluate their potential as reactive media for permeable reactive barriers (PRBs) to remove aqueous trichloroethylene (TCE) from groundwater. Batch tests indicated that the slags exhibit varying degrees of
Young Kim et al.
Biotechnology and bioengineering, 80(5), 498-508 (2002-10-02)
Batch kinetic and inhibition studies were performed for the aerobic cometabolism of 1,1,1-trichloroethane (1,1,1-TCA), 1,1-dichloroethylene (1,1-DCE), and 1,1-dichloroethane (1,1-DCA) by a butane-grown mixed culture. These chlorinated aliphatic hydrocarbons (CAHs) are often found together as cocontaminants in groundwater. The maximum degradation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service