Skip to Content
Merck
All Photos(1)

Key Documents

24246

Supelco

Carboxen®-1010 PLOT Capillary GC Column

L × I.D. 30 m × 0.32 mm, average thickness 15 μm

Synonym(s):

Carboxen® 1010 PLOT Column, 30m x 0.32mm

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115710

material

fused silica

Agency

ASTM® D3612-96

description

PLOT = Porous Layer Open Tubular

manufacturer/tradename

Carboxen®

parameter

≤25-250 °C temperature (isothermal or programmed)

average thickness

15 μm

df

15 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.32 mm

matrix active group

Carbon molecular sieve phase

application(s)

chemicals and industrial polymers
food and beverages

column type

capillary PLOT

Looking for similar products? Visit Product Comparison Guide

General description

Application: This column is ideal for the separation of all major components in permanent gas (helium, hydrogen, oxygen, nitrogen, carbon monoxide, methane, and carbon dioxide) and light hydrocarbons (C2-C3) in the same analysis. It is the only column commercially available that is able to separate all major components in permanent gas. This column can also separate oxygen from nitrogen at subambient temperatures.
USP Code: None
Phase:
  • Carbon molecular sieve
Temp. Limits:
  • Subambient to 250 °C (isothermal or programmed)
Porous-layer open-tubular (PLOT) columns are a powerful tool suitable in gas chromatography for analysis of fixed gases, light hydrocarbons and volatile solvents. They are mostly preferred over traditional columns.

Application

Gas chromatograph (GC), equipped with a 1010 Carboxen® GC Plot capillary column and a thermal conductivity detector (TCD) was also used for the chemical analyses of the composition of product gas in the batch enrichment assays, during bioprospecting thermophilic microorganisms derived from Icelandic hot springs which are efficient producers of hydrogen and ethanol.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bioprospecting Thermophilic Microorganisms from Icelandic Hot Springs for Hydrogen and Ethanol Production.
Koskinen P
Energy and Fuels, 22 (1), 134-140 (2007)
Porous layer open-tubular capillary columns: preparations, applications and future directions.
Journal of Chromatography A, 842 (1), 115-142 (1999)
Sebastien Allard et al.
Journal of chromatography. A, 1238, 15-21 (2012-04-13)
Trihalomethanes are predominantly formed during disinfection of water via reactions of the oxidant with natural organic matter. Even though chlorinated and brominated trihalomethanes are the most widespread organic contaminants in drinking water, when iodide is present in raw water iodinated
Dawn M Chapman et al.
Journal of agricultural and food chemistry, 52(17), 5431-5435 (2004-08-19)
A rapid and automated solid phase microextraction (SPME) stable isotope dilution gas chromatography/mass spectrometry (GC-MS) method for 2-methoxy-3-isobutylpyrazine (MIBP) quantification in red wine was developed. Wines with 30% (w/v) NaCl and 2-methoxy-(2)H(3)-3-isobutylpyrazine internal standard were sampled with a 2 cm
Lucia Giordano et al.
Journal of chromatography. A, 1017(1-2), 141-149 (2003-10-31)
A new method was developed for the determination of 2-furfural (2-F) and 5-methylfurfural (5-MF), two products of Maillard reaction in vinegar, with head-space solid-phase microextraction (HS-SPME) coupled to gas chromatography-mass spectrometry (GC-MS). A divinylbenzene (DVB)/carboxen (CAR)/polydimethylsiloxane (PDMS) fibre was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service