Skip to Content
Merck
All Photos(1)

Key Documents

21341-U

Supelco

Silica Gel

22% H₂SO₄ Coated, pkg of 100 g bulk packing

Synonym(s):

Sulfuric Acid Coated Silica Gel

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
13111000
NACRES:
NB.21

Product Name

22% H2SO4 Coated Silica Gel, pkg of 100 g bulk packing

Agency

suitable for EPA 1613

Quality Level

packaging

pkg of 100 g bulk packing

technique(s)

solid phase extraction (SPE): suitable

particle size

60-200 mesh

application(s)

food and beverages

General description

The treated silica gels, which can be mainly used in pre-packed multi-layer silica gel columns, are offered in bulk packages. These are potential options for customizing the desired columns to more efficiently clean very dirty samples, or to prepare shorter columns whenever the samples are relatively clean, e.g. drinking water.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service