21318-U
Silica Gel
2% KOH Coated, pkg of 100 g bulk packing
Synonym(s):
KOH Coated Silica Gel
About This Item
Recommended Products
Product Name
2% KOH Coated Silica Gel, pkg of 100 g bulk packing
Agency
suitable for EPA 1613
Quality Level
form
solid
packaging
pkg of 100 g bulk packing
technique(s)
solid phase extraction (SPE): suitable
particle size
60-200 mesh
application(s)
food and beverages
Related Categories
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service