Skip to Content
Merck
All Photos(1)

Key Documents

20276

Supelco

Amberlite XAD4

20-60 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

form

solid

autoignition temp.

800 °F

packaging

pkg of 100 g

technique(s)

HPLC: suitable
LPLC: suitable

surface area

750 m2/g

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

20-60 mesh

pore size

~0.98 mL/g pore volume
100 Å mean pore size

density

1.02 g/mL (true wet)(lit.)
1.08 g/mL (skeletal)(lit.)

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite XAD4 is a nonpolar absorbent used for stripping and concentration of organic compounds. Bacillus subtilis immobilized on Amberlite XAD-4 has been investigated as new biosorbent in trace metal determination.
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

  • Amberlite XAD4 was used in rapid fractionation technique.
  • Amberlite XAD4 was used as sorbent for HPLC analysis.
Polyaromatic adsorbent for small hydrophobic compounds, surfactants, pharmaceutical manufacturing, phenols, chlorinated organics, pesticide removal and recovery, and organic removal from aqueous food streams.

Other Notes

Two particle sizes are available:
Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4
Mesh 20-50 is Cat. No. 06444

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Characterization and applications of Amberlite XAD-4 in preparative liquid chromatography
Pietrzyk, D. J., and J. D. Stodola
Analytical Chemistry, 53, 1822-1828 (1981)
Passively capturing air data on more volatile chemicals.
Kellyn Betts
Environmental science & technology, 43(4), 978-978 (2009-03-27)
A S Ghatbandhe et al.
Journal of environmental science & engineering, 50(2), 163-168 (2009-03-20)
Adsorption equilibrium, kinetics and thermodynamics of 2,4-dichlorophenol (2,4-DCP), one of the most commonly used chlorophenol, onto bituminous coal based Filtrasorb-400 grade granular activated carbon, were studied in aqueous solution in a batch system with respect to temperature. Uptake capacity of
Muhammad Afzal Kamboh et al.
Journal of hazardous materials, 172(1), 234-239 (2009-07-31)
The present study describes a novel synthetic method for the immobilization of calix[4]arene (II) onto the surface of modified Amberlite XAD-4 resin (4), which does not require the derivatization of calixarene moiety. The novel calix[4]arene based resin (C4 resin) 5
Siva Sarathy et al.
Water research, 44(14), 4087-4096 (2010-06-18)
The advanced oxidation process utilizing ultraviolet and hydrogen peroxide (UV/H(2)O(2)) is currently applied in commercial drinking water applications for the removal of various organic pollutants. Natural organic matter (NOM) present in the source water can also be oxidized and undergo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service