Skip to Content
Merck
All Photos(1)

Key Documents

00268

Supelco

Dextran

analytical standard, for GPC, 1,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

form

powder or crystals

mol wt

Mn ~1,010
Mp ~1,080
Mw ~1,270

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.26

application(s)

food and beverages

format

neat

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is a polysaccharide. Dextran fluids behave as colloids and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

Application


  • Dextran for drug delivery: Dextran′s role as a biopolymer in drug delivery systems is underscored by its capacity to enhance drug solubility and stability, facilitating controlled release mechanisms crucial for targeted therapy applications (Wagai et al., 2024).

  • Dextran as a blood plasma expander: Dextran serves as a blood plasma expander in medical treatments, effectively restoring blood volume in patients undergoing surgery or suffering from shock, due to its high molecular weight and osmotic properties (Kohashi et al., 2024).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshikazu Isomura et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(25), 10209-10220 (2013-06-21)
It is widely accepted that dorsal striatum neurons participate in either the direct pathway (expressing dopamine D1 receptors) or the indirect pathway (expressing D2 receptors), controlling voluntary movements in an antagonistically balancing manner. The D1- and D2-expressing neurons are activated
Krystal Nizar et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(19), 8411-8422 (2013-05-10)
Calcium-dependent release of vasoactive gliotransmitters is widely assumed to trigger vasodilation associated with rapid increases in neuronal activity. Inconsistent with this hypothesis, intact stimulus-induced vasodilation was observed in inositol 1,4,5-triphosphate (IP3) type-2 receptor (R2) knock-out (KO) mice, in which the
Kelli G Sharp et al.
Experimental neurology, 257, 186-204 (2014-04-22)
As part of the NIH "Facilities of Research Excellence-Spinal Cord Injury" project to support independent replication, we repeated key parts of a study reporting robust engraftment of neural stem cells (NSCs) treated with growth factors after complete spinal cord transection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service