Skip to Content
Merck
All Photos(1)

Key Documents

00270

Supelco

Dextran

analytical standard, for GPC, 12,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

form

powder or crystals

mol wt

Mn ~8,110
Mp ~9,890
Mw ~11,600

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.43

application(s)

food and beverages

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

Application


  • Application in cancer therapies:The study discusses the conjugation of dextran with therapeutic enzymes to enhance their stability and efficacy, underscoring its significance in developing more effective cancer therapies (El-Sayed et al., 2023).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Peritoneum, 324-324 (2012)
An Evidence-based Approach
Alexander, Mary; Corrigan, Ann;
Infusion Nursing, 239-239 (2009)
Dmitri Simberg et al.
Biomaterials, 30(23-24), 3926-3933 (2009-04-28)
In order to understand the role of plasma proteins in the rapid liver clearance of dextran-coated superparamagnetic iron oxide (SPIO) in vivo, we analyzed the full repertoire of SPIO-binding blood proteins using novel two-dimensional differential mass spectrometry approach. The identified
Ana Luisa Miranda-Vilela et al.
Journal of biomedical nanotechnology, 9(7), 1261-1271 (2013-08-06)
This work aimed to test a dextran-functionalized magnetic fluid (DexMF) sample in mediating magnetohyperthermia to treat an advanced clinical Ehrlich-solid-tumor, to verify the effects of oral antioxidant administration of pequi-oil on this treatment and to investigate the potential of these
David Needham et al.
Faraday discussions, 161, 515-534 (2013-06-29)
This paper describes how we have used material science, physical chemistry, and some luck, to design a new thermal-sensitive liposome (the low temperature sensitive liposome (LTSL)) that responds at clinically attainable hyperthermic temperatures releasing its drug in a matter of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service