Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1286

Sigma-Aldrich

Anti-VE-Cadherin Antibody, clone 1O12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O12, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 87.53 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O12 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects VE-Cadherin. It targets an epitope within 15 amino acids from the extracellular domain in the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the extracellular domain in the C-terminal half of human VE-Cadherin.

Application

Quality Control Testing

Evaluated by Western Blotting in HUVEC lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected VE-Cadherin (CD144) in human umbilical vein endothelial cell (HUVEC) lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected VE-Cadherin in human brain tissue lysate.

Affinity Binding Assay: A representative lot of this antibody bound VE-Cadherin peptide with a KD of 9 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected VE-Cadherin in human testis and human cerebral cortex tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected VE-Cadherin in one million human umbilical vein endothelial cells (HUVEC).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected VE-Cadherin in human umbilical vein endothelial cells (HUVEC).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-VE-Cadherin, clone 1O12 ZooMAb®, Cat. No. ZRB1286, is a recombinant Rabbit monoclonal antibody that targets VE-Cadherin and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Cadherin-5 (UniProt: P33151; also known as 7B4 antigen, Vascular endothelial cadherin, VE-cadherin, CD144) is encoded by the CDH5 gene (Gene ID: 1003) in human. Cadherins are calcium-dependent cell adhesion proteins that preferentially interact with themselves in a homophilic manner in connecting cells and contribute to the sorting of heterogeneous cell types. They play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. VE-Cadherin is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-25) and a propeptide (aa 26-47) that are subsequently cleaved off to generate the mature form that contain an extracellular domain (aa 48-599) with five cadherin repeats, a transmembrane domain (aa 600-620), and a cytoplasmic domain ( aa 621-784). Three calcium ions are usually bound at the interface of each cadherin domain and rigidify the connections, providing a strong curvature to the full-length ectodomains. VE-cadherin is mainly expressed in endothelial tissue and in the brain and plays a pivotal role in endothelium integrity and vascular permeability. It associates with a-catenin forming a link to the cytoskeleton. It acts in concert with Krev interaction trapped protein 1 (KRIT1) and membrane palmitoylated protein 5 (MPP5) to establish and maintain correct endothelial cell polarity and vascular lumen. Amino acids region 621-660 is required for its interaction with MPP5. Two isoforms of Cadherin 5 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service