Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1277

Sigma-Aldrich

Anti-F4/80 Antibody, clone 2F10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F10, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~97.68 kDa
observed mol wt ~97 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2F10 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects F4/80 (Adhesion G protein-coupled receptor E1). It targets an epitope within 15 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human F4/80 (Adhesion G protein-coupled receptor E1).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected F4/80 in Jurkat cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected F4/80 in Raji, NIH3T3, and human spleen tissue lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected F4/80 in Jurkat cells.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected F4/80 in one million Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound F4/80 peptide with a KD of 2.0 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: 1:100 from a representative lot detected F4/80 in human spleen and human bone marrow tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-F4/80, clone 2F10 ZooMAb®, Cat. No. ZRB1277, is a recombinant Rabbit monoclonal antibody that targets F4/80 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Adhesion G protein-coupled receptor E1 (UniProt: Q14246; also known as EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EMR1 hormone receptor, F4/80) is encoded by the ADGRE1 (also known as EMR1, TM7LN3) gene (Gene ID: 2015) in human. F4/80 is multi-pass membrane glycoprotein of the G-protein coupled receptor 2 family that contains four extracellular domains, seven transmembrane domains, and four cytoplasmic domains. It also has six EGF-like domains in the N-terminal region followed by a serine/threonine-rich stalk region. F4/80 is involved in cell adhesion and in cell-cell interactions involving the immune system. F4/80 is reported to be a pan-macrophage marker mouse to detect the distribution and localization of tissue macrophages and to examine the phenotypic heterogeneity and activation state of resident macrophages. Although not required for normal macrophage development in murine species, F4/80 is shown to be essential for the induction of efferent CD8+ regulatory T-cells, which is needed for peripheral tolerance. In human F4/80 expression is restricted to eosinophils. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service