Skip to Content
Merck
All Photos(1)

Key Documents

SAB5500103

Sigma-Aldrich

Anti-F4/80 antibody, Rabbit monoclonal

clone SP115, recombinant, expressed in proprietary host, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP115, monoclonal

species reactivity

mouse (tested)

species reactivity (predicted by homology)

rat

technique(s)

immunohistochemistry: 1:100

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ADGRE1(2015)

General description

EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1) is a human homolog of F4/80, an eosinophil-specific receptor. It is abundantly expressed on leukocytes. F4/80 is a member of the GPCR superfamily. It has N-terminal EGF (epidermal growth factor)-like domains bound to a seven-span transmembrane (7TM) moiety through a mucin-like spacer domain. This gene is located on human chromosome 19p13.
Mouse EGF-like module-containing mucin-like hormone receptor-like 1 (EMR1) or cell surface glycoprotein F4/80 is a cell surface glycoprotein and a member of the EGF TM7 family of proteins. It shares 68% overall amino acid identity with human EGF-module-containing-mucin-like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ regulatory T cells.

Immunogen

Synthetic peptide derived from N-terminus of mouse F4/80 protein.

Application

Anti-F4/80 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

In liquid bone marrow cultures, F4/80/EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1) acts as a maturation marker for mononuclear phagocytes. EMR1 can be used as a target molecule in the treatment of individuals suffering from squamous cell carcinoma and eosinophilic disorders.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EMR1, the human homolog of F4/80, is an eosinophil-specific receptor
Hamann J, et al.
European Journal of Immunology, 37(10), 2797-2802 (2007)
Kelly L Harris et al.
Oncotarget, 7(20), 28947-28960 (2016-03-10)
Consumption of Western diet (WD), contaminated with environmental toxicants, has been implicated as one of the risk factors for sporadic colon cancer. Our earlier studies using a mouse model revealed that compared to unsaturated dietary fat, the saturated dietary fat
The mononuclear phagocyte system of the mouse defined by immunohistochemical localization of antigen F4/80. Relationship between macrophages, Langerhans cells, reticular cells, and dendritic cells in lymphoid and hematopoietic organs.
Hume DA, et al.
The Journal of Experimental Medicine, 158(5), 1522-1536 (1983)
Western diet enhances benzo(a)pyrene-induced colon tumorigenesis in a polyposis in rat coli (PIRC) rat model of colon cancer
Harris KL, et al.
Oncotarget, 7(20), 28947-28960 (2016)
Human EMR2, a novel EGF-TM7 molecule on chromosome 19p13.1, is closely related to CD97
Lin HH, et al.
Genomics, 67(2), 188-200 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service