Z743327
ContainerSEAL® sealing tape
W × L 0.75 in. × 108 ft, yellow
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech CSEAL-75Y
W × L
0.75 in. × 108 ft
color
yellow
Related Categories
General description
ContainerSEAL is a sealing tape designed for the vast range of bottles and containers used in laboratory applications. Its unique elastomeric characteristics allow it to stretch and conform around the irregular perimeters of containers lids and covers to form a tight seal. Container Seal will block water vapor loss, reduce the rate of gas exchange and prevent the entry of contaminants.
Ideally sized (0.75" wide), the tape can be easily removed from most surfaces without leaving an adhesive residue. Available in a variety of colors.
Ideally sized (0.75" wide), the tape can be easily removed from most surfaces without leaving an adhesive residue. Available in a variety of colors.
Legal Information
ContainerSEAL is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)