If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
Z104388
PTFE sealing tape
W 1/2 in.
Synonym(s):
Thread Sealing PTFE Tape
About This Item
Recommended Products
material
TFE
sterility
non-sterile
packaging
pack of 3 rolls
manufacturer/tradename
Sigma-Aldrich
W
1/2 in.
roll size
520 in. (13.2 m)
thickness × width × L
3 mil × 1/2 in. × 520 in.
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Gas cylinder extraction setups
- Microfluidic device plumbing setups
- Carrier gas connections for gas chromatography
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
Qual è la temperatura massimo di esercizio di questo prodotto (PTFE sealing tape)? Può andare in autoclave (121°C per 20 minuti)
1 answer-
Yes, this product may be autoclaved. The material can be autoclaved at 121°C for 20 minutes. The maximum operating temperature of polytetrafluoroethylene (PTFE or Teflon) is approximately 500°F. If heated at or below this temperature, the material does not undergo thermal degradation. Please note the thermal decomposition products are highly dependent upon the combustion conditions. Polytetrafluoroethylene can emit toxic fumes when heated to temperatures exceeding 700°F.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service