Z742405
PELCO® Sliding Latch Microslide Storage Boxes
white, Holds 20 x slides 76x26 mm
Synonym(s):
slide storage
All Photos(1)
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
Recommended Products
material
acrylonitrile-butadiene-styrene (ABS)
white
feature
Holds 20 x slides 76x26 mm
packaging
pack of 12 ea
manufacturer/tradename
Ted Pella 2306-1-CS
L × W × H
104 mm (4.10 in.) × 94 mm (3.70 in.) × 32 mm (1.26 in.)
Related Categories
General description
Exceptional slide storage containers providing maximum protection for your microscope slides.
Features and Benefits
- Numbered index sheet in lid for quick identification
- Foam liner
- Stackable
- ABS-durable/impact resistant
- Hinged lid
- Unique sliding compression latch
Legal Information
PELCO is a registered trademark of Ted Pella, Inc
Choose from one of the most recent versions:
Certificates of Analysis (COA)