Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PELCO
®
Sliding Latch Microslide Storage Boxes
Products
Product Category
boxes (21)
Manufacturer
Height (mm)
Length (in)
Length (mm)
Color
Package Size
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z742390
PELCO
®
Sliding Latch Microslide Storage Boxes,
white, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742397
PELCO
®
Sliding Latch Microslide Storage Boxes,
pink, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742411
PELCO
®
Sliding Latch Microslide Storage Boxes,
black
Expand
View Pricing
Z742404
PELCO
®
Sliding Latch Microslide Storage Boxes,
pink, Holds 50 x slides 76x26 mm
Expand
View Pricing
Z742409
PELCO
®
Sliding Latch Microslide Storage Boxes,
blue, Holds 20 x slides 76x26 mm
Expand
View Pricing
Z742393
PELCO
®
Sliding Latch Microslide Storage Boxes,
green, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742395
PELCO
®
Sliding Latch Microslide Storage Boxes,
yellow, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742396
PELCO
®
Sliding Latch Microslide Storage Boxes,
black, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742405
PELCO
®
Sliding Latch Microslide Storage Boxes,
white, Holds 20 x slides 76x26 mm
Expand
View Pricing
Z742394
PELCO
®
Sliding Latch Microslide Storage Boxes,
blue, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742400
PELCO
®
Sliding Latch Microslide Storage Boxes,
green, Holds 50 x slides 76x26 mm
Expand
View Pricing
Z742401
PELCO
®
Sliding Latch Microslide Storage Boxes,
blue, Holds 50 x slides 76x26 mm
Expand
View Pricing
Z742408
PELCO
®
Sliding Latch Microslide Storage Boxes,
green, Holds 20 x slides 76x26 mm
Expand
View Pricing
Z742402
PELCO
®
Sliding Latch Microslide Storage Boxes,
yellow, Holds 50 x slides 76x26 mm
Expand
View Pricing
Z742410
PELCO
®
Sliding Latch Microslide Storage Boxes,
yellow, Holds 20 x slides 76x26 mm
Expand
View Pricing
Z742403
PELCO
®
Sliding Latch Microslide Storage Boxes,
black, Holds 50 x slides 76x26 mm
Expand
View Pricing
Z742407
PELCO
®
Sliding Latch Microslide Storage Boxes,
red, Holds 20 x slides 76x26 mm
Expand
View Pricing
Z742391
PELCO
®
Sliding Latch Microslide Storage Boxes,
red, Holds 100 x slides 76x26 mm
Expand
View Pricing
Z742399
PELCO
®
Sliding Latch Microslide Storage Boxes,
red, Holds 50 x slides 76x26 mm
Expand
View Pricing
Z742412
PELCO
®
Sliding Latch Microslide Storage Boxes,
pink, Holds 20 x slides 76x26 mm
Expand
View Pricing
Page 1 of 2
Page 1 of 2