Skip to Content
Merck
All Photos(1)

Documents

Z741768

Sigma-Aldrich

Sigma-Aldrich® Dual Run and Blot System

Synonym(s):

SDS-PAGE equipment, electrophoresis and blotting equipment

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

feature

Fits both 10x10 and 8x10 cassettes

manufacturer/tradename

Sigma-Aldrich

General description

Sigma Aldrich′s Dual Run & Blot system provides you with a gel tank with unsurpassed functionality. The core module hinges open to accept gel cassettes or buffer dam and then simply clips in place to provide a completely sealed cathode chamber. This allows RunBuffer to be added and samples to be loaded outside of the tank. In addition, Safety Cover removal could not be easier, with two protruding posts to push against, it can be slid off smoothly!

The Dual Run & Blot system utilises a unique, patent pending, gradient electric filed for blotting which provides greatest potential for the largest proteins and less for the smaller proteins, giving the most uniform blotting ever!

Features and Benefits

  • Fits both 10x10 and 8x10 cassettes
  • Colours centre core to provide maximum contrast with loading buffer for easy loading
  • Excellent sealing and no leakage from cathode to anode
  • Can be cooled in place to reduce heat effects such as smiling in frowning
  • Gradient field for blotting providing more uniform transfer. Ideal for high quality wet blotting
  • Excellent value for money

Components

  • Lower Reservoir Assembly (x1)
  • Safety Cover with Attached DC Leads (x1)
  • Electrophoresis Core Modue (x1)
  • Blotting Cassettes (x2)
  • Sponge Pads (x6)
  • Buffer Dam (x1)
  • Cooling Blocks (x2)

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service