BM-100
Auto2D® 2-D Electrophoresis Device
automates 2D gel electrophoresis
Synonym(s):
2D Electrophoresis, 2DE, IEF, SDS-PAGE
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41100000
NACRES:
NC.01
Recommended Products
product line
Auto2D®
technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
western blot: suitable
storage temp.
room temp
General description
The Auto2D® 2-D Electrophoresis Device automates two-dimensional gel electrophoresis. The applied sample enters the first-dimension gel passively, separating proteins by their isoelectric points. After equilibration, the first-dimension gel is set onto a horizontal SDS-PAGE gel, which resolves proteins by molecular weight.
Two-dimensional gel electrophoresis has generally been regarded as difficult to perform and time-consuming, requireing advanced user training while offering low reproducibility and high inter-operator variability.
Two-dimensional gel electrophoresis has generally been regarded as difficult to perform and time-consuming, requireing advanced user training while offering low reproducibility and high inter-operator variability.
Application
- Differential proteomic analysis
- Post-translational modification
- Protein Isoforms
- Western Blotting
Features and Benefits
Auto2D® 2-D Electrophoresis Device is user-friendly and enable two-dimensional electrophoresis in 1-2 hours for faster, more reproducible results.
- Fully automated to eliminate tedious handling steps
- Fast results in 1-2 hours
- High reproducibility
- Easy to use, with no advanced training required
- Lower inter-operator variability
Storage and Stability
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Legal Information
Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service