Z708550
Square weighing boats
size M
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.71
Recommended Products
material
polystyrene
square
size
M
packaging
pack of 500 ea
manufacturer/tradename
Heathrow Scientific HS1421B
L × W × H
89 mm × 89 mm × 25 mm
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Polystyrene weighing boats with a smooth surface to provide accurate pour-outs with minimal sample loss. Shallow shape with a wide, flat bottom to resist tipping. Rounded corners simplify transfer. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases. May be used with microwave moisture analyzers. Resist temperatures to 200 °F (93 °C).
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service