Skip to Content
Merck
All Photos(1)

Key Documents

Z0417

Sigma-Aldrich

β-Zearalanol

~98% (HPLC)

Synonym(s):

2,4-Dihydroxy-6-(6β,10-dihydroxyundecyl]benzoic acid μ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O5
CAS Number:
Molecular Weight:
322.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

Quality Level

Assay

~98% (HPLC)

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

availability

not available in Canada

solubility

methanol: 19.60-20.40 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@@H](O)CCCCCc2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H26O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,14,19-21H,2-9H2,1H3/t12-,14-/m0/s1

InChI key

DWTTZBARDOXEAM-JSGCOSHPSA-N

Gene Information

rat ... Ar(24208)

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zearalenones: characterization of the estrogenic potencies and receptor interactions of a series of fungal beta-resorcylic acid lactones.
B S Katzenellenbogen et al.
Endocrinology, 105(1), 33-40 (1979-07-01)
B H Migdalof et al.
Xenobiotica; the fate of foreign compounds in biological systems, 13(4), 209-221 (1983-04-01)
The disposition of [3H]zeranol has been studied in the female Wistar rat, New Zealand rabbit, beagle dog, rhesus monkey and man. The blood elimination half-life of total radioactivity in rabbit was 26 h, monkey 18 h and man 22 h.
Erika Pfeiffer et al.
Molecular nutrition & food research, 54(10), 1468-1476 (2010-04-17)
Glucuronidation constitutes an important pathway in the phase II metabolism of the mycotoxin zearalenone (ZEN) and the growth promotor α-zearalanol (α-ZAL, zeranol), but the enzymology of their formation is yet unknown. In the present study, ZEN, α-ZAL and four of
Alberto Sánchez Arribas et al.
Electrophoresis, 30(3), 499-506 (2009-01-22)
A simple analytical scheme for the screening and quantification of zearalenone and its metabolites, alpha-zearalenol and beta-zearalenol, is reported. Extracts from maize flour samples were collected by supercritical fluid extraction and afterwards, they were analyzed by CE with amperometric detection.
A Laganá et al.
Rapid communications in mass spectrometry : RCM, 15(4), 304-310 (2001-02-27)
An analytical procedure that enables routine analysis for trace determination of six anabolic macrocyclic lactones (zearalenone, alpha- and beta-zearalenol, zearalanone, zeranol, and taleranol) in sewage treatment plant (STP) samples has been developed. The method uses solid-phase extraction, followed by high-performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service