Skip to Content
Merck

Skip To

D4628

Diethylstilbestrol

≥99% (HPLC), Synthetic estrogen, powder

Synonym(s):

(E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, DES, Stilbestrol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
EC Number:
200-278-5
MDL number:
Beilstein/REAXYS Number:
2056095
Assay:
≥99% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Diethylstilbestrol, ≥99% (HPLC)

Quality Segment

assay

≥99% (HPLC)

form

powder

SMILES string

CC\C(c1ccc(O)cc1)=C(\CC)c2ccc(O)cc2

InChI

1S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20H,3-4H2,1-2H3/b18-17+

InChI key

RGLYKWWBQGJZGM-ISLYRVAYSA-N

Gene Information

Application

Diethylstilbestrol has been used:
  • to evaluate the estrogenic activity of diethylstilbestrol by quantitating the expression levels of endogenous estrogen-regulated marker genes
  • to evaluate the estrogenic, androgenic and toxicity responses in bioluminescent yeast bioreporter assay (BLYES)
  • to detect its effect on the proliferation and tyrosinase activity of melanocytes

Biochem/physiol Actions

Diethylstilbestrol is a synthetic estrogen with carcinogenic properties. Causes renal clear-cell carcinoma in Syrian hamster. In humans it causes increased risk of breast cancer, clear cell adenocarcinoma (CCA) of the vagina and cervix, and reproductive anomalies. Used in the treatment of prostate cancer to block the production of testosterone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
462071195001BP859
description

≥99% (HPLC)

description

VETRANAL®, analytical standard

description

United States Pharmacopeia (USP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

form

powder

form

-

form

-

form

powder

assay

≥99% (HPLC)

assay

-

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

Gene Information

human ... ESR1(2099), ESR2(2100), ESRRG(2104)
mouse ... Esr1(13982)
rat ... Ar(24208), Esr1(24890)

Gene Information

human ... ESR1(2099)

Gene Information

human ... ESR1(2099)

Gene Information

human ... ESR1(2099)


Still not finding the right product?

Explore all of our products under Diethylstilbestrol


Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

PPE (personal protective equipment)

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters