Skip to Content
Merck
All Photos(1)

Key Documents

X4879

Sigma-Aldrich

Xaliproden hydrochloride

≥97% (HPLC), solid

Synonym(s):

1,2,3,6-Tetrahydro-1-[2-(2-naphthalenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]pyridine hydrochloride, SR-57746A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22F3N · HCl
CAS Number:
Molecular Weight:
417.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: ≥5 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

Cl.FC(F)(F)c1cccc(c1)C2=CCN(CCc3ccc4ccccc4c3)CC2

InChI

1S/C24H22F3N.ClH/c25-24(26,27)23-7-3-6-22(17-23)20-11-14-28(15-12-20)13-10-18-8-9-19-4-1-2-5-21(19)16-18;/h1-9,11,16-17H,10,12-15H2;1H

InChI key

WVHBEIJGAINUBW-UHFFFAOYSA-N

Gene Information

human ... HTR1A(3350)

Biochem/physiol Actions

5-HT1A serotonin receptor agonist; neuroprotectant

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Terranova et al.
Neuroscience letters, 213(2), 79-82 (1996-08-02)
Different putative toxic amyloid beta (A beta) peptides, beta (1-42), beta (1-40) and beta (25-35), were infused (0.75, 1.5 or 3 nmol) in the rat medial septum. Memory deficits were then investigated using the social recognition test. A significant amnesia
Highlights From: the 2006 American Society of Clinical Oncology Gastrointestinal Cancers Symposium San Francisco, CA, January 2006.
Tara Beers Gibson et al.
Clinical colorectal cancer, 5(6), 398-402 (2006-04-26)
A Appert-Collin et al.
International journal of immunopathology and pharmacology, 17(2), 157-164 (2004-06-03)
Compounds possessing neurotrophic properties may represent a possible treatment for neurodegenerative disorders such as amyotrophic lateral sclerosis. Xaliproden (SR57746A), an orally-active non-peptide compound, which has been found to exhibit neurotrophic effects in vitro and in vivo, increased the lifespan and
Sherry Wolf et al.
European journal of cancer (Oxford, England : 1990), 44(11), 1507-1515 (2008-06-24)
Chemotherapy-induced peripheral neuropathy (CIPN) is a major dose limiting side effect of many commonly used chemotherapeutic agents, including platinum drugs, taxanes, epothilones and vinca alkaloids, and also newer agents such as bortezomib and lenolidamide. Symptom control studies have been conducted
What we have learned from the Xaliproden Sanofi-aventis trials.
P Douillet et al.
The journal of nutrition, health & aging, 13(4), 365-366 (2009-03-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service