Skip to Content
Merck
All Photos(6)

Key Documents

X3375

Sigma-Aldrich

Xylitol

≥99% (GC)

Synonym(s):

Xylite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]3CH2OH
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720523
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (GC)

form

crystalline
crystals or chunks
powder

sweetness

1 × sucrose

color

colorless

mp

94-97 °C (lit.)

solubility

water: 642g/l at 25 °C (77 °F )

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+

InChI key

HEBKCHPVOIAQTA-SCDXWVJYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xylitol is a naturally occurring five carbon sugar alcohol, equivalent to sucrose in sweetness. Xylitol finds applications in the preparation of confectionaries, chewing gum, toothpaste and mouthwashes. Xylitol is a low-energy sweetener with insulin independent metabolism, making it a promising alternative for sugar in diabetic patients. Xylitol is a natural anticaries agent used in the treatment of dental caries, as it is not utilized by cariogenic bacteria creates a starvation effect on them. Xylitol prevents otitis and upper respiratory tract infections. Commercially, microorganisms like bacteria, fungi and yeasts produce xylitol by fermentation.

Application

Xylitol has been used:
  • as a standard for high performance liquid chromatography
  • in analysing antibacterial activity on oral biofilms
  • for thermodynamic data and densities measurement

Biochem/physiol Actions

A sugar alcohol sweetener detectable by humans. Produced from hemicellulose hydrolysate fermentation.

Other Notes

To gain a comprehensive understanding of our extensive range of sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular strategies for enhancing microbial production of xylitol
Pal S, et al.
Process Biochemistry (Oxford, United Kingdom), 51(7), 809-819 (2016)
Synergistic effect of xylitol and ursolic acid combination on oral biofilms
Zou Y, et al.
Restorative dentistry & endodontics, 39(4), 288-295 (2014)
Culture parameters affecting xylitol production by Debaryomyces hansenii immobilized in alginate beads
Perez-Bibbins B, et al.
Process Biochemistry (Oxford, United Kingdom), 48(3), 387-397 (2013)
Apparent molar volumes and apparent molar heat capacities of aqueous adonitol, dulcitol, glycerol, meso-erythritol, myo-inositol, D-sorbitol, and xylitol at temperatures from (278.15 to 368.15) K and at the pressure 0.35 MPa
Blodgett MB, et al.
The Journal of Chemical Thermodynamics, 39(4), 627-644 (2007)
Eun Joong Oh et al.
Metabolic engineering, 15, 226-234 (2012-10-30)
As Saccharomyces cerevisiae cannot utilize xylose as a carbon source, expression of XYL1 coding for xylose reductase (XR) from Scheffersomyces (Pichia) stipitis enabled production of xylitol from xylose with a high yield. However, insufficient supply of NAD(P)H for XR and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service