Skip to Content
Merck
All Photos(1)

Documents

W507930

Sigma-Aldrich

Xylitol

Synonym(s):

Xylite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]3CH2OH
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720523
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder or crystals

mp

94-97 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+

InChI key

HEBKCHPVOIAQTA-SCDXWVJYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiet A Ly et al.
Pediatric dentistry, 28(2), 154-163 (2006-05-20)
The purpose of this report was to provide an overview of xylitol and other polyol sweeteners and dental caries for clinicians and to discuss current applications for dental practice and potential community-based public health interventions. Xylitol, like other polyol sweeteners
Tom Birger Granström et al.
Applied microbiology and biotechnology, 74(2), 277-281 (2007-01-12)
The rare sugar xylitol is a five-carbon polyol (pentitol) that has beneficial health effects. Xylitol has global markets and, therefore, it represents an alternative to current dominant sweeteners. The research on microbial reduction of D-xylose to xylitol has been focused
Eun Joong Oh et al.
Metabolic engineering, 15, 226-234 (2012-10-30)
As Saccharomyces cerevisiae cannot utilize xylose as a carbon source, expression of XYL1 coding for xylose reductase (XR) from Scheffersomyces (Pichia) stipitis enabled production of xylitol from xylose with a high yield. However, insufficient supply of NAD(P)H for XR and
L Zhan et al.
Journal of dental research, 91(7 Suppl), 85S-90S (2012-09-21)
The aim of the study was to investigate the efficacy of the use of xylitol-containing tooth-wipes in preventing dental caries in young children. In a double-blinded randomized controlled clinical trial, 44 mothers with active caries and their 6- to 35-month-old
Lilian Raquel Hickert et al.
Bioresource technology, 143, 112-116 (2013-06-25)
Co-fermentation and simultaneous saccharification of rice hull hydrolysate (RHH) were investigated for the production of ethanol and xylitol by Saccharomyces cerevisiae, Spathaspora arborariae, or the combination of both. In bioreactor cultures under oxygen limitation, S. cerevisiae was capable of metabolizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service