Skip to Content
Merck
All Photos(3)

Key Documents

WH0005337M1

Sigma-Aldrich

Monoclonal Anti-PLD1 antibody produced in mouse

clone 2F3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-phospholipase D1, phophatidylcholine-specific

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLD1(5337)

General description

Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis (Hammond et al., 1995 [PubMed 8530346]).[supplied by OMIM

Immunogen

PLD1 (NP_002653, 965 a.a. ~ 1074 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GYLDDPSEDIQDPVSDKFFKEVWVSTAARNATIYDKVFRCLPNDEVHNLIQLRDFINKPVLAKEDPIRAEEELKKIRGFLVQFPFYFLSEESLLPSVGTKEAIVPMEVWT

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service