Skip to Content
Merck
All Photos(2)

Documents

WH0007442M1

Sigma-Aldrich

Monoclonal Anti-TRPV1 antibody produced in mouse

clone 1F5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Trpv1 Antibody, Trpv1 Antibody - Monoclonal Anti-TRPV1 antibody produced in mouse, Anti-DKFZp434K0220, Anti-VR1, Anti-transient receptor potential cation channel, subfamily V, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPV1(7442)

General description

The gene encoding transient receptor potential cation channel subfamily V member 1 (TRPV1) is mapped to human chromosome 17p13. It is a non-selective cation channel. The protein is expressed on airway nerve fibers. It is also strongly expressed in sensory neurons.

Immunogen

TRPV1 (NP_542437, 21 a.a. ~ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
CPDPLDGDPNSRPPPAKPQLSTAKSRTRLFGKGDSEEAFPVDCPHEEGELDSCPTITVSPVITIQRPGDGPTGARLLSQDSVAASTEKTLRLYDRRSIFEAVAQ

Biochem/physiol Actions

Transient receptor potential cation channel subfamily V member 1 (TRPV1) is a capsaicin receptor. It participates in pain perception. TRPV1 also modulates afferent signals and bronchoconstriction. In the brain, it regulates neuronal function, motor behaviour and neuroinflammation. Capsaicin plays an important role in the activation of TRPV1.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of transient receptor potential vanilloid 1 in the modulation of airway smooth muscle tone and calcium handling.
Yocum GT
American Journal of Physiology. Lung Cellular and Molecular Physiology (2017)
Gitit Kra et al.
Animals : an open access journal from MDPI, 12(6) (2022-03-26)
Environmental heat load (HL) adversely affects the performance of dairy cows. The endocannabinoid system (ECS) regulates metabolism and the stress response, thus we hypothesized that HL may affect the ECS of dairy cows. Our objective was to determine the levels
The 57 kb deletion in cystinosis patients extends into TRPV1 causing dysregulation of transcription in peripheral blood mononuclear cells.
Freed KA
Journal of medical Genetics (2011)
J R Daddam et al.
Journal of dairy science (2023-08-29)
We examined the effects of a supplement of plant polyphenols extracts of green tea, capsicum and fenugreek, and electrolytes [(Na+, K+), AXT; Axion ThermoPlus, CCPA, France] during summer heat load on production, welfare, and oxidative stress proteins in adipose tissue
TRPV1 on astrocytes rescues nigral dopamine neurons in Parkinson's disease via CNTF.
Nam JH
Brain (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service