Skip to Content
Merck
All Photos(1)

Documents

W108

Sigma-Aldrich

WAY-100635 maleate salt

powder

Synonym(s):

N-[2-[4-(2-Methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinylcyclohexanecarboxamide maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34N4O2 · C4H4O4
CAS Number:
Molecular Weight:
538.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

color

white

solubility

H2O: 25 mg/mL

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.COc1ccccc1N2CCN(CC2)CCN(C(=O)C3CCCCC3)c4ccccn4

InChI

1S/C25H34N4O2.C4H4O4/c1-31-23-12-6-5-11-22(23)28-18-15-27(16-19-28)17-20-29(24-13-7-8-14-26-24)25(30)21-9-3-2-4-10-21;5-3(6)1-2-4(7)8/h5-8,11-14,21H,2-4,9-10,15-20H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

XIGAHNVCEFUYOV-BTJKTKAUSA-N

Gene Information

human ... HTR1A(3350)

Application

WAY-100635 maleate salt has been used as a 5-HT1A serotonin receptor antagonist:
  • to study whether the behavior of the cleaner wrasse is regulated by serotonin activity
  • to block activity of 3,4 methylenedioxymethamphetamine (MDMA) used to induce oxytocin release in male Wistar rats to increase their social behavior
  • to study the role of serotonin during metamorphosis of the ascidian Phallusia mammillata larvae

Biochem/physiol Actions

WAY-100635 maleate salt is a highly selective 5-HT1A serotonin receptor antagonist. It has an ability to inhibit brexpiprazole, an antipsychotic drug that regulates serotonin-dopamine activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naozumi Araragi et al.
Frontiers in pharmacology, 4, 97-97 (2013-08-13)
Firing activity of serotonin (5-HT) neurons in the dorsal raphe nucleus (DRN) is controlled by inhibitory somatodendritic 5-HT1A autoreceptors. This autoinhibitory mechanism is implicated in the etiology of disorders of emotion regulation, such as anxiety disorders and depression, as well
Mehdi Sekssaoui et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 49(4), 747-756 (2024-01-12)
Major depressive disorder (MDD) is one of the most disabling psychiatric disorders in the world. First-line treatments such as selective serotonin reuptake inhibitors (SSRIs) still have many limitations, including a resistance to treatment in 30% of patients and a delayed
Effects of brexpiprazole, a novel serotonin-dopamine activity modulator, on phencyclidine-induced cognitive deficits in mice: a role for serotonin 5-HT1A receptors
Yoshimi N, et al.
Pharmacology, Biochemistry, and Behavior, 124, 245-249 (2014)
H Gozlan et al.
European journal of pharmacology, 288(2), 173-186 (1995-01-16)
The tritiated derivative of the novel silent 5-HT1A receptor antagonist WAY 100635 [N-(2-(4-(2-methoxyphenyl)-1-piperazinyl)ethyl)-N-(2-pyridinyl) cyclohexane carboxamide] was tested as a potential radioligand of 5-HT1A receptors in the rat brain. Binding assays with membranes from various brain regions showed that [3H]WAY 100635
Elizabeth G Pitts et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 42(10), 1962-1971 (2017-04-21)
3,4-Methylenedioxymethamphetamine (MDMA) increases sociality in humans and animals. Release of serotonin (5-HT) is thought to have an important role in the increase in social behaviors, but the mechanisms underlying these effects are poorly understood. Despite the advantages of nonhuman primate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service