Skip to Content
Merck
All Photos(3)

Key Documents

V7000

Sigma-Aldrich

Snake venom from Crotalus atrox (Western Diamondback Rattlesnake)

Synonym(s):

Crotalus atrox Venom, Rattlesnake Venom, Western Diamondback Rattlesnake Venom

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352108
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

storage temp.

−20°C

Application

Snake venom from Crotalus atrox (Western Diamondback Rattlesnake) has been used:
  • in phosphodiesterase 3 (PDE3) and phosphodiesterase 4 (PDE4) inhibition assay[1]
  • in the reaction mixture for PDE3B assay[2]
  • in PDE assay[3]
  • to convert all nucleoside- 5′-monophosphate to guanosine and inorganic phosphate[4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-induced phosphorylation and activation of cyclic nucleotide phosphodiesterase 3B by the serine-threonine kinase Akt
Kitamura T, et al.
Molecular and Cellular Biology, 19(9), 6286-6296 (1999)
Photoreceptor phosphodiesterase: interaction of inhibitory gamma subunit and cyclic GMP with specific binding sites on catalytic subunits
Artemyev N O, et al.
Methods, 14(1), 93-104 (1998)
Augmentation of lipolysis in adipocytes from fed rats, but not from starved rats, by inhibition of rolipram-sensitive phosphodiesterase 4
Nakamura J, et al.
Archives of Biochemistry and Biophysics, 425(1), 106-114 (2004)
Hunseung Yoo et al.
Journal of ethnopharmacology, 206, 298-305 (2017-05-17)
Sophora tonkinensis (Leguminosae, ST) is a traditional herbal plant in Korea and China. Its roots and rhizomes have been used to dissipate heat, to clear toxic material and to treat acute pharyngolaryngeal infections and sore throats. In this study, we

Questions

  1. Does this item come in a liquid form? What should I resuspend it in? Elaine

    1 answer
    1. A similar product, A9378, is listed under the same EC number (232-564-0) and is provided as an aqueous suspension at 5mg/mL.
      This would indicate V7000 can be dissolved in water as well at this concentration.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service