Skip to Content
Merck
All Photos(1)

Key Documents

V9014

Sigma-Aldrich

Anti-VMAT2 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SLC18A2, Anti-SVAT, Anti-SVMT, Anti-Solute carrier family 18 (vesicular monoamine), member 2, Anti-VAT2, Anti-VMAT2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:250
western blot: 1:500-1:1,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Vesicular monoamine transporter 2 (VMAT2) , also known as solute carrier family 18 member A2 (SLC18A2), is a transmembrane protein that acts at nerve terminals. It is expressed in platelets and in central, peripheral and enteric neurons. The gene encoding VMAT2 consists of 16 exons and is mapped to human chromosome 10q25.

Immunogen

synthetic peptide corresponding an internal sequence of human VMAT2. This sequence is identical in mouse and rat.

Application

Anti-VMAT2 antibody produced in rabbit has also been used in immunofluorescence.
Anti-VMAT2 antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

Vesicular monoamine transporter 2 (VMAT2) modulates the release of neurotransmitters like dopamine, serotonin and γ-amino butyric acid. It has a role in the accumulation of neurotransmitters in synaptic vesicles to prevent toxicity. VMAT2 has been linked to tardive dyskinesia and may be associated with Parkinson′s disease.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Age-dependent dopamine transporter dysfunction and Serine129 phospho-α-synuclein overload in G2019S LRRK2 mice
Longo F
Acta Neuropathologica Communications, 5, 22-22 (2017)
Norepinephrine transporter expression is inversely associated with glycaemic indices: a pilot study in metabolically diverse persons with overweight and obesity.
Straznicky NE
Obesity science & practice, 2, 13-23 (2016)
Vesicular monoamine transporter, type 2 (VMAT2) expression as it compares to insulin and pancreatic polypeptide in the head, body and tail of the human pancreas.
Freeby M
Islets, 4, 393-397 (2012)
Analysis of vesicular monoamine transporter 2 polymorphisms in Parkinson's disease.
Brighina L
Neurobiology of Aging, 34, e9-13 (2013)
Decreased striatal vesicular monoamine transporter 2 (VMAT2) expression in a type 1 diabetic rat model: A longitudinal study using micro-PET/CT
Jiang D, et al.
Nuclear Medicine and Biology, 82, 89-95 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service