Skip to Content
Merck
All Photos(1)

Documents

V8261

Sigma-Aldrich

(±)-Vigabatrin

Synonym(s):

(R,S)-4-Amino-5-hexenoic acid, (±)-γ-Vinyl-GABA, (±)-4-Aminohexenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

originator

Lundbeck

SMILES string

NC(CCC(O)=O)C=C

InChI

1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)

InChI key

PJDFLNIOAUIZSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Irreversible GABA transaminase inhibitor. Increases intracellular concentration of GABA in nerve endings; possesses antiepileptic activity.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABA Transporters, GABAA Receptors and Glutamate/GABA Synthesis and Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Lundbeck. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Doszyn et al.
iScience, 27(6), 110149-110149 (2024-07-01)
Mechanistic target of rapamycin complex 1 (mTORC1) is an integration hub for extracellular and intracellular signals necessary for brain development. Hyperactive mTORC1 is found in autism spectrum disorder (ASD) characterized by atypical reactivity to sensory stimuli, among other symptoms. In
R S Fisher
Brain research. Brain research reviews, 14(3), 245-278 (1989-07-01)
The study of mechanisms of the epilepsies requires employment of animal models. Choice of a model system depends upon several factors, including the question to be studied, the type of epilepsy to be modelled, familiarity and convenience. Over 50 models
The potential value of a negative finding: an illustrative example.
Kenneth Silverman et al.
JAMA psychiatry, 70(6), 571-572 (2013-04-12)
Nicolas Froger et al.
Advances in experimental medicine and biology, 775, 69-83 (2013-02-09)
Retinal ganglion cells (RGCs) are spiking neurons, which send visual information to the brain, through the optic nerve. RGC degeneration occurs in retinal diseases, either as a primary process or secondary to photoreceptor loss. Mechanisms involved in this neuronal degeneration
Jose I Juncosa et al.
Bioorganic & medicinal chemistry, 21(4), 903-911 (2013-01-12)
We have synthesized three analogues of 4-amino-5-fluorohexanoic acids as potential inactivators of γ-aminobutyric acid aminotransferase (GABA-AT), which were designed to combine the potency of their shorter chain analogue, 4-amino-5-fluoropentanoic acid (AFPA), with the greater enzyme selectivity of the antiepileptic vigabatrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service