Skip to Content
Merck
All Photos(1)

Key Documents

V4014

Sigma-Aldrich

Anti-V5-Cy3 antibody, Mouse monoclonal

purified immunoglobulin, ~1 mg/mL, clone V5-10

Synonym(s):

Monoclonal Anti-V5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

CY3 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

V5-10, monoclonal

concentration

~1 mg/mL

technique(s)

immunocytochemistry: 2.0-4.0 μg/mL

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-V5 (mouse IgG1 isotype) is derived from the V5 hybridoma, produced by fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to amino acid residues GKPIPNPLLGLDST (95-108) of the P/V proteins of the Paramyxovirus SV5, conjugated to KLH. The conjugate is then extensively dialyzed to remove unbound Cy3 fluorophore. Monoclonal Anti-V5, Cy3 conjugate recognizes the V5 tag sequence on V5 tagged fusion proteins by immunocytochemistry.

Specificity

Recognizes V5 Tag (GKPIPNPLLGLDST) fusion proteins

Application

Many recombinant proteins have been engineered to express a short sequence derived from the V5 molecule known as V5-tag.[1][2] This tag facilitates the detection, isolation and purification of the proteins.[3] Monoclonal antibodies reacting specifically with V5 and conjugated to Cy3 may be useful in various immunotechniques, and also to identify the expression of a V5 tagged fusion protein in situ by immunofluorescence.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Legal Information

Cy3 is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Southern et al.
The Journal of general virology, 72 ( Pt 7), 1551-1557 (1991-07-01)
Two canine isolates of simian virus 5 (SV5), termed CPI+ and CPI-, were examined for their ability to react with a bank of monoclonal antibodies (MAbs) that had been previously raised against a human isolate of SV5. CPI- virus was
Dharmendra K Singh et al.
Molecular plant-microbe interactions : MPMI, 27(1), 7-17 (2013-09-21)
Protein kinase-driven phosphorylation constitutes the core of cellular signaling. Kinase components of signal transduction pathways are often targeted for inactivation by pathogens. The study of kinases and immune signal transduction in the model crop tomato (Solanum lycopersicum) would benefit from
C Dunn et al.
Journal of immunological methods, 224(1-2), 141-150 (1999-06-05)
The monoclonal antibody (mAb) SV5-Pk is used widely in a variety of procedures to detect recombinant proteins tagged with the Pk tag, a 14 amino acid sequence derived from the P and V proteins of the paramyxovirus Simian Virus 5.
S M Thomas et al.
Cell, 54(6), 891-902 (1988-09-09)
The "P" gene of the paramyxovirus SV5 encodes two known proteins, P (Mr approximately equal to 44,000) and V (Mr approximately equal to 24,000). The complete nucleotide sequence of the "P" gene has been obtained and is found to contain
Suena Ji et al.
Nature communications, 9(1), 3508-3508 (2018-08-31)
Regulated secretion is an essential process where molecules destined for export are directed to membranous secretory granules, where they undergo packaging and maturation. Here, we identify a gene (pgant9) that influences the structure and shape of secretory granules within the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service