Skip to Content
Merck
All Photos(1)

Documents

S6639

Sigma-Aldrich

SSC Buffer 20× Concentrate

for Northern and Southern blotting, solution

Synonym(s):

20X SSC Buffer, SSC hybridization buffer, Saline-sodium citrate buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

grade

for molecular biology

Quality Level

description

Saline-Sodium Citrate buffer, made with ultrapure water

sterility

0.2 μm filtered

form

solution

application(s)

diagnostic assay manufacturing

foreign activity

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
RNAse, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

SSC (Saline Sodium Citrate) Buffer 20x liquid is a standard reagent in Southern and Northern hybridization procedures. It is used to control stringency of wash buffer for the washing steps after hybridization.

Application

SSC Buffer 20× Concentrate has been used
  • as a hybridization buffer on in situ hybridization human-specific repetitive Alu sequence
  • as a washing buffer for membrane for Southern hybridization, dot blot of mice genomic DNA and northern blotting of microRNA
  • as an incubation buffer in fluorescence in situ hybridization (FISH) of L1 retrotransposon

Features and Benefits

  • Convenient ready-to-use liquid
  • Multi-purpose use in hybridizations

Components

SSC Buffer 20x contains 3M NaCl in 0.3M sodium citrate (pH 7.0).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Gong Liu et al.
Methods (San Diego, Calif.), 44(1), 22-30 (2007-12-26)
After 12 years from its first application, microarray technology has become the reference technique to monitor gene expression of thousands of genes in the same experiment. In the past few years an increasing amount of evidence showed the importance of
Chronic hypoxia facilitates Alzheimer's disease through demethylation of gamma-secretase by downregulating DNA methyltransferase 3b
Liu H, et al.
Alzheimers Dement., 12(2), 130-143 (2016)
M Böttger et al.
Nucleic acids research, 9(20), 5253-5268 (1981-10-24)
Complexes of histones H1 with superhelical SV40 DNA obtained by direct mixing were studied in 0.1 SSC buffer corresponding to 0.02 M Na+. Depending on the molar input ratio H1/DNA three classes of sedimenting species were observed: (1) a component
Michaela Asp et al.
Scientific reports, 7(1), 12941-12941 (2017-10-13)
Heart failure is a major health problem linked to poor quality of life and high mortality rates. Hence, novel biomarkers, such as fetal marker genes with low expression levels, could potentially differentiate disease states in order to improve therapy. In
L1 retrotransposon expression in circulating tumor cells
Papasotiriou I, et al.
Testing, 12(2), e0171466-e0171466 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service