Skip to Content
Merck
All Photos(1)

Documents

V1628

Sigma-Aldrich

Verrucarol

Synonym(s):

4β,15-Dihydroxy-12,13-epoxytrichothec-9-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O4
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

storage temp.

−20°C

SMILES string

[H][C@]12C[C@@H](O)C(C)([C@@]3(CO)CCC(C)=C[C@@]3([H])O1)C24CO4

InChI

1S/C15H22O4/c1-9-3-4-14(7-16)11(5-9)19-12-6-10(17)13(14,2)15(12)8-18-15/h5,10-12,16-17H,3-4,6-8H2,1-2H3/t10-,11-,12-,13?,14-,15?/m1/s1

InChI key

ZSRVBNXAPSQDFY-CXOCQXMISA-N

Biochem/physiol Actions

Trichothecene mycotoxin that is a component of toxic mold. Esters of verrucarol have anti-tumor activity.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Barel et al.
Journal of pharmaceutical sciences, 79(6), 548-551 (1990-06-01)
Verrucarol is a simple trichothecene which is structurally related to T-2 and HT-2 toxins. Several macrocyclic trichothecenes which are ester derivatives of verrucarol possess antitumor activity. The pharmacokinetics of verrucarol has been studied in eight dogs following iv and oral
T L Brasel et al.
Applied and environmental microbiology, 71(1), 114-122 (2005-01-11)
Highly respirable particles (diameter, <1 microm) constitute the majority of particulate matter found in indoor air. It is hypothesized that these particles serve as carriers for toxic compounds, specifically the compounds produced by molds in water-damaged buildings. The presence of
B Harrach et al.
Applied and environmental microbiology, 41(6), 1428-1432 (1981-06-01)
A strain of Stachybotrys atra isolated from a field case of stachybotryotoxicosis in Hungary was cultured in Hungary. All of the compounds toxic to brine shrimp were separated from the culture extract by solvent partition, column chromatography, and preparative thin-layer
Mirko Peitzsch et al.
Journal of environmental monitoring : JEM, 14(3), 908-915 (2012-01-31)
We compared the efficiency of some commercially available products and methods used for remediation of mould-contaminated building materials. Samples of gypsum board and pinewood were artificially contaminated with toxin-producing isolates of Stachybotrys chartarum and Aspergillus versicolor, respectively, then, ten different
T Tuomi et al.
Applied and environmental microbiology, 66(5), 1899-1904 (2000-05-02)
We analyzed 79 bulk samples of moldy interior finishes from Finnish buildings with moisture problems for 17 mycotoxins, as well as for fungi that could be isolated using one medium and one set of growth conditions. We found the aflatoxin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service